Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032061.2 | NP_114450.1 | 17 | 2800 | ||
XM_006247431.2 | XP_006247493.1 | 17 | 2822 | |||
XM_008768122.1 | XP_008766344.1 | 17 | 2812 | |||
XM_008768123.1 | XP_008766345.1 | 17 | 2615 | |||
XM_008768124.1 | XP_008766346.1 | 16 | 2579 | |||
GenBank mRNA | AF000114.1 | - | 17 | 2788 | ||
U87224.1 | - | 17 | 2800 |