Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053369.1 | NP_445821.1 | 14 | 1480 | ||
GenBank mRNA | AF149284.1 | - | 14 | 1480 | ||
FQ221525.1 | - | 1231 | ||||
FQ226283.1 | - | 5 | 619 | |||
FQ227003.1 | - | 2 | 225 | |||
FQ232404.1 | - | 2 | 224 | |||
FQ233045.1 | - | 10 | 1112 | |||
U09228.1 | - | 8 | 887 |