Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105737.1 | NP_001099207.1 | 21, 22 | 3517 | ||
XM_008763910.1 | XP_008762132.1 | 21, 22 | 3337 |