Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS27 SIRNA Assays ›
          Gene Symbol:
          Rps27 LOC100362987 LOC100364561 LOC100911728 LOC102554672 Slfn14
          Gene Name:
          ribosomal protein S27
          ribosomal protein S27-like
          40S ribosomal protein S27-like
          40S ribosomal protein S27-like
          40S ribosomal protein S27-like
          schlafen family member 14
          Gene Aliases:
          -
          -
          -
          -
          -
          LRRGT00163
          Chromosome Location:
          Chr.2: 189572175 - 189573280 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 10737527 - 10737830 on Build Rnor_6.0
          Chromosome Location:
          Chr.12: 40669050 - 40669751 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 89218835 - 89219225 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 128079033 - 128079561 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 70493340 - 70504800 on Build Rnor_6.0
          UniGene ID:
          Rn.144623
          -
          -
          -
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s137235

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053597.3 NP_446049.1 2, 3 156
          GenBank mRNA AF184893.1 - 2, 3 150
          BC061539.1 - 160 6918361
          FQ209846.1 - 2, 3 156
          FQ210151.1 - 2, 3 155
          FQ217607.1 - 2, 3 158
          FQ217624.1 - 2, 3 156
          FQ218228.1 - 2, 3 157
          FQ218581.1 - 2, 3 156
          FQ221131.1 - 2, 3 156
          FQ221294.1 - 2, 3 156
          FQ221645.1 - 2, 3 155
          FQ221664.1 - 2, 3 156
          FQ221723.1 - 2, 3 155
          FQ221727.1 - 2, 3 153
          FQ221743.1 - 2, 3 156
          FQ221809.1 - 2, 3 156
          FQ221911.1 - 2, 3 156
          FQ221975.1 - 2, 3 157
          FQ222049.1 - 2, 3 156
          FQ222054.1 - 2, 3 157
          FQ222270.1 - 2, 3 157
          FQ222348.1 - 3, 4 273
          FQ222848.1 - 2, 3 156
          FQ222865.1 - 2, 3 156
          FQ222911.1 - 2, 3 156
          FQ223072.1 - 2, 3 156
          FQ223291.1 - 2, 3 155
          FQ223549.1 - 2, 3 156
          FQ223731.1 - 2, 3 157
          FQ223862.1 - 2, 3 156
          FQ224473.1 - 2, 3 156
          FQ228507.1 - 2, 3 157
          FQ228599.1 - 2, 3 156
          FQ228823.1 - 2, 3 156
          FQ229526.1 - 2, 3 156
          Gene Symbol: LOC100362987
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002730094.4 XP_002730140.1 1 123
          Gene Symbol: LOC100364561
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006249411.2 XP_006249473.1 1 259
          Gene Symbol: LOC100911728
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003751271.3 XP_003751319.1 1 205
          Gene Symbol: LOC102554672
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008763965.1 XP_008762187.1 2 195
          Gene Symbol: Slfn14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY539914.1 - 123

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001030, L19739, BC002658, AV763942, U57847, and others.
          This siRNA also targets the following transcripts in Mouse: NM_027015, NM_001190258, BC048352, BC055693, BC027649, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          structural constituent of ribosome
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0