Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057186.2 | NP_476534.1 | 4 | 515 | ||
GenBank mRNA | AF095449.1 | - | 4 | 515 | ||
FQ212781.1 | - | 4 | 495 |