Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CITED2 SIRNA Assays ›
          Gene Symbol:
          Cited2
          Gene Name:
          Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2
          Gene Aliases:
          Mrg1
          Chromosome Location:
          Chr.1: 12823363 - 12825806 on Build Rnor_6.0
          UniGene ID:
          Rn.31765
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s137634

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cited2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053698.2 NP_446150.1 2 593
          GenBank mRNA AF361476.1 - 2 598
          BC087005.1 - 2 593 7191504

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001168389, NM_006079, NM_001168388, AF109161, U65093, and others.
          This siRNA also targets the following transcripts in Mouse: NM_010828, AK133791, BC057126, AK131664, Y15163, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          vasculogenesis
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          kidney development
          trophectodermal cell differentiation
          neural tube formation
          neural tube closure
          liver development
          embryonic placenta development
          vasculature development
          heart looping
          morphogenesis of an epithelium
          lens morphogenesis in camera-type eye
          hematopoietic progenitor cell differentiation
          leukocyte differentiation
          outflow tract morphogenesis
          regulation of organ formation
          endocardial cushion development
          ventricular septum development
          regulation of transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          determination of left/right symmetry
          central nervous system development
          peripheral nervous system development
          heart development
          sex determination
          cell aging
          cell proliferation
          male gonad development
          response to mechanical stimulus
          positive regulation of gene expression
          negative regulation of gene expression
          cranial nerve morphogenesis
          positive regulation of cell-cell adhesion
          adrenal gland development
          negative regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          granulocyte differentiation
          response to fluid shear stress
          positive regulation of peroxisome proliferator activated receptor signaling pathway
          adrenal cortex formation
          skeletal muscle cell differentiation
          negative regulation of apoptotic process
          response to estrogen
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          spleen development
          thymus development
          embryonic camera-type eye morphogenesis
          erythrocyte development
          embryonic process involved in female pregnancy
          bone morphogenesis
          cardiac septum morphogenesis
          ventricular septum morphogenesis
          embryonic heart tube left/right pattern formation
          pulmonary artery morphogenesis
          cardiac neural crest cell development involved in heart development
          determination of heart left/right asymmetry
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          left/right axis specification
          cellular response to growth factor stimulus
          nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          regulation of nucleic acid-templated transcription
          positive regulation of male gonad development
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          RNA polymerase II transcription corepressor activity
          molecular_function
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          transcription corepressor activity
          histone acetyltransferase binding
          SMAD binding
          LBD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline