Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229825.2 | XP_006229887.1 | 9 | 1817 | ||
XM_006229830.2 | XP_006229892.1 | 9 | 1806 | |||
XM_006229831.2 | XP_006229893.1 | 9 | 1629 | |||
XM_006229832.2 | XP_006229894.1 | 9 | 1617 | |||
XM_006229834.2 | XP_006229896.1 | 8 | 1585 | |||
XM_006229835.2 | XP_006229897.1 | 6 | 1968 | |||
XM_006229837.2 | XP_006229899.1 | 8 | 1400 | |||
XM_006229838.2 | XP_006229900.1 | 8 | 1388 | |||
XM_008759662.1 | XP_008757884.1 | 6 | 1368 | |||
XM_008759663.1 | XP_008757885.1 | 6 | 1909 | |||
GenBank mRNA | AB236144.1 | - | 7 | 875 | ||
FQ230423.1 | - | 2 | 104 | |||
U57715.1 | - | 8 | 1380 |