Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053903.1 | NP_446355.1 | 4 | 537 | ||
GenBank mRNA | U69279.1 | - | 4 | 537 |