Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
Tpm3
Gene Name:
tropomyosin 3
Gene Aliases:
TM30nm, Tpm5
Chromosome Location:
Chr.2: 189423539 - 189451345 on Build Rnor_6.0
UniGene ID:
Rn.37575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s138842
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301285.1
NP_001288214.1
4
493
NM_001301286.1
NP_001288215.1
3
317
NM_057208.2
NP_476556.2
3
317
NM_173111.1
NP_775134.1
3
359
XM_006232568.2
XP_006232630.1
4
513
XM_006232569.2
XP_006232631.1
4
513
XM_006232570.2
XP_006232632.1
4
513
XM_006232571.2
XP_006232633.1
4
513
XM_006232572.2
XP_006232634.1
4
513
XM_006232573.2
XP_006232635.1
4
513
XM_006232575.2
XP_006232637.1
4
513
XM_006232576.2
XP_006232638.1
4
513
XM_006232577.2
XP_006232639.1
4
513
XM_006232580.2
XP_006232642.1
3
417
XM_006232581.2
XP_006232643.1
3
418
XM_006232583.2
XP_006232645.1
3
421
XM_006232584.2
XP_006232646.1
3
419
XM_008761124.1
XP_008759346.1
3
417
XM_008761125.1
XP_008759347.1
3
518
XM_008761126.1
XP_008759348.1
2
284
GenBank mRNA
FQ214881.1
-
4
491
FQ215580.1
-
4
493
FQ215752.1
-
4
493
FQ216429.1
-
4
492
FQ228755.1
-
3
367
L24775.1
-
3
317
L24776.1
-
317
L24777.1
-
3
317
S82383.1
-
3
336
X72859.1
-
3
359
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001271764, NM_001253738, NM_001253740, NM_022314, AK166836, and others.
Gene Ontology Categories:
Function(s)
Process(es)
brain development
molecular_function
actin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0