Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057209.1 | NP_476557.1 | 7 | 1287 | ||
GenBank mRNA | J03886.1 | - | 7 | 1287 |