Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133532.2 | NP_598216.2 | 1 | 778 | ||
XM_006243915.2 | XP_006243977.1 | 2 | 965 | |||
GenBank mRNA | AF432872.2 | - | 1 | 778 |
Set Membership: |
![]() |