Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100639.1 | NP_001094109.1 | 11 | 1202 | ||
XM_008769591.1 | XP_008767813.1 | 11 | 3328 | |||
XM_008769592.1 | XP_008767814.1 | 11 | 1463 | |||
XM_008769593.1 | XP_008767815.1 | 13 | 1662 | |||
XM_008769594.1 | XP_008767816.1 | 12 | 1564 | |||
XM_008769595.1 | XP_008767817.1 | 13 | 1597 | |||
XM_008769596.1 | XP_008767818.1 | 13 | 3772 | |||
XM_008769597.1 | XP_008767819.1 | 12 | 3674 | |||
XM_008769598.1 | XP_008767820.1 | 11 | 3573 | |||
XM_008769599.1 | XP_008767821.1 | 11 | 1226 | |||
GenBank mRNA | D12978.1 | - | 3 | 279 | ||
U17013.1 | - | 6 | 792 |