Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134332.2 | NP_599159.1 | 4 | 502 | ||
GenBank mRNA | FQ219852.1 | - | 4 | 565 | ||
FQ219889.1 | - | 4 | 565 | |||
FQ219947.1 | - | 4 | 563 | |||
FQ219966.1 | - | 4 | 563 | |||
FQ219998.1 | - | 4 | 567 | |||
FQ220005.1 | - | 4 | 565 | |||
FQ220014.1 | - | 4 | 565 | |||
FQ220035.1 | - | 4 | 565 | |||
FQ220062.1 | - | 4 | 567 | |||
FQ220064.1 | - | 4 | 565 | |||
FQ220079.1 | - | 4 | 563 | |||
FQ220128.1 | - | 4 | 562 | |||
FQ220142.1 | - | 4 | 565 | |||
FQ220223.1 | - | 4 | 565 | |||
FQ220228.1 | - | 4 | 555 | |||
FQ220364.1 | - | 4 | 565 | |||
FQ220426.1 | - | 4 | 565 | |||
FQ220453.1 | - | 4 | 565 | |||
FQ220464.1 | - | 4 | 565 | |||
FQ220466.1 | - | 4 | 565 | |||
FQ220480.1 | - | 4 | 567 | |||
FQ220512.1 | - | 4 | 565 | |||
FQ220556.1 | - | 4 | 565 | |||
FQ220562.1 | - | 4 | 565 | |||
FQ220618.1 | - | 4 | 565 | |||
FQ220648.1 | - | 4 | 565 | |||
FQ220735.1 | - | 4 | 565 | |||
FQ220749.1 | - | 4 | 564 | |||
FQ220751.1 | - | 4 | 563 | |||
FQ220777.1 | - | 4 | 565 | |||
FQ220856.1 | - | 4 | 564 | |||
FQ220878.1 | - | 4 | 564 | |||
FQ220928.1 | - | 4 | 567 | |||
FQ221390.1 | - | 4 | 565 | |||
FQ221473.1 | - | 4 | 565 | |||
FQ221896.1 | - | 2 | 288 | |||
FQ221940.1 | - | 4 | 565 | |||
FQ223110.1 | - | 4 | 565 | |||
FQ223477.1 | - | 4 | 564 | |||
FQ228922.1 | - | 4 | 567 | |||
FQ229160.1 | - | 4 | 565 | |||
FQ229161.1 | - | 4 | 565 | |||
FQ229162.1 | - | 4 | 565 | |||
FQ229181.1 | - | 4 | 567 | |||
FQ229215.1 | - | 4 | 565 | |||
FQ229235.1 | - | 4 | 569 | |||
FQ229289.1 | - | 4 | 565 | |||
FQ229298.1 | - | 4 | 565 | |||
FQ229311.1 | - | 4 | 565 | |||
FQ229314.1 | - | 4 | 565 | |||
FQ229333.1 | - | 4 | 565 | |||
FQ229395.1 | - | 4 | 565 | |||
FQ229439.1 | - | 4 | 563 | |||
FQ229447.1 | - | 4 | 563 | |||
FQ229452.1 | - | 4 | 565 | |||
FQ229454.1 | - | 4 | 565 | |||
FQ229468.1 | - | 4 | 565 | |||
FQ229486.1 | - | 4 | 565 | |||
FQ229502.1 | - | 4 | 565 | |||
FQ229509.1 | - | 4 | 565 | |||
FQ229543.1 | - | 4 | 565 | |||
FQ229636.1 | - | 4 | 565 | |||
FQ229685.1 | - | 4 | 565 | |||
FQ229690.1 | - | 4 | 609 | |||
FQ229764.1 | - | 4 | 566 | |||
FQ229772.1 | - | 4 | 563 | |||
FQ229825.1 | - | 4 | 565 | |||
FQ229868.1 | - | 4 | 596 | |||
FQ229912.1 | - | 4 | 565 | |||
FQ229913.1 | - | 4 | 563 | |||
FQ229994.1 | - | 4 | 567 | |||
FQ230028.1 | - | 4 | 567 | |||
FQ230087.1 | - | 4 | 565 | |||
FQ230100.1 | - | 4 | 565 | |||
FQ230157.1 | - | 4 | 567 | |||
FQ230165.1 | - | 4 | 565 | |||
U57062.1 | - | 4 | 503 |
Gene Symbol: LOC502003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770722.1 | XP_008768944.1 | 2 | 296 |
Gene Symbol: LOC691695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751488.3 | XP_003751536.2 | 4 | 623 | ||
GenBank mRNA | FQ219891.1 | - | 4 | 565 | ||
FQ229899.1 | - | 4 | 565 |