Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134414.2 | NP_599241.2 | 10 | 1201 | ||
XM_006253891.1 | XP_006253953.1 | 11 | 1273 | |||
XM_006253893.2 | XP_006253955.1 | 10 | 1157 | |||
XM_006253895.2 | XP_006253957.1 | 9 | 1085 | |||
GenBank mRNA | AF032666.1 | - | 10 | 1139 | ||
CK482593.1 | - | 6 | 650 |
Gene Symbol: LOC103689971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253876.2 | XP_006253938.1 | 1273 | |||
XM_006253878.2 | XP_006253940.1 | 1156 | ||||
XM_006253880.2 | XP_006253942.1 | 1084 | ||||
XM_006253881.2 | XP_006253943.1 | 1273 | ||||
XM_008771607.1 | XP_008769829.1 | 1201 |