Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100912262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249222.2 | XP_006249284.1 | 4 | 1069 | ||
XM_006249223.2 | XP_006249285.1 | 4 | 1069 | |||
XM_006249224.2 | XP_006249286.1 | 4 | 1069 | |||
XM_006249225.2 | XP_006249287.1 | 4 | 1069 | |||
XM_006249226.2 | XP_006249288.1 | 4 | 1069 | |||
XM_006249227.2 | XP_006249289.1 | 4 | 1069 |
Gene Symbol: Gtf2ird1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001504.2 | NP_001001504.1 | 5 | 599 | ||
XM_006249102.2 | XP_006249164.1 | 5 | 768 | |||
XM_006249103.2 | XP_006249165.1 | 5 | 801 | |||
XM_006249107.2 | XP_006249169.1 | 5 | 765 | |||
XM_008769115.1 | XP_008767337.1 | 5 | 801 | |||
XM_008769116.1 | XP_008767338.1 | 5 | 801 | |||
XM_008769117.1 | XP_008767339.1 | 5 | 801 | |||
XM_008769118.1 | XP_008767340.1 | 5 | 801 | |||
XM_008769119.1 | XP_008767341.1 | 5 | 800 | |||
XM_008769120.1 | XP_008767342.1 | 5 | 800 | |||
XM_008769121.1 | XP_008767343.1 | 5 | 802 | |||
XM_008769122.1 | XP_008767344.1 | 5 | 802 | |||
XM_008769123.1 | XP_008767345.1 | 5 | 802 | |||
XM_008769124.1 | XP_008767346.1 | 5 | 802 | |||
GenBank mRNA | AF547388.1 | - | 5 | 599 | ||
AF547389.1 | - | 5 | 599 | |||
FQ217037.1 | - | 5 | 660 |