Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145677.1 | NP_663710.1 | 1 | 153 | ||
XM_006233935.2 | XP_006233997.1 | 1 | 1490 | |||
GenBank mRNA | AY043169.1 | - | 1 | 153 | ||
FQ211394.1 | - | 1 | 169 |