Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153297.1 | NP_695209.1 | 5 | 591 | ||
XM_006234901.2 | XP_006234963.1 | 5 | 541 | |||
XM_008762146.1 | XP_008760368.1 | 5 | 541 | |||
GenBank mRNA | AB081072.1 | - | 5 | 591 | ||
BC061864.1 | - | 5 | 512 | 5597874 | ||
FQ210124.1 | - | 5 | 515 |