Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153308.4 | NP_695220.4 | 4 | 718 | ||
GenBank mRNA | BC062074.1 | - | 4 | 688 | 5599428 | |
S61973.1 | - | 4 | 668 |