Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172329.1 | NP_758832.1 | 3 | 705 | ||
XM_006244179.2 | XP_006244241.1 | 3 | 1511 | |||
GenBank mRNA | AF458780.1 | - | 3 | 705 | ||
BC091115.1 | - | 3 | 719 | 7381553 |
Set Membership: |
![]() |