Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 16 | 3406 | ||
NM_005426.2 | NP_005417.1 | 17 | 3538 | |||
XM_011544268.1 | XP_011542570.1 | 15 | 3267 | |||
XM_011544269.2 | XP_011542571.1 | 13 | 2925 | |||
XM_017002220.1 | XP_016857709.1 | 17 | 3547 | |||
GenBank mRNA | AJ318888.2 | - | 16 | 3351 | ||
AK294432.1 | - | 16 | 3258 | |||
AK294654.1 | - | 6 | 1342 | |||
AK294964.1 | - | 9 | 2426 | |||
AK295378.1 | - | 10 | 2130 | |||
AK316016.1 | - | 9 | 2328 | |||
AK316103.1 | - | 9 | 2306 | |||
AL357233.1 | - | 4 | 580 | |||
BC040247.1 | - | 14 | 2882 | 4454266 | ||
BC058918.1 | - | 16 | 3364 | 4537206 | ||
U09582.1 | - | 5 | 1301 | |||
U58334.1 | - | 17 | 3483 |