Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPD52L1 SIRNA Assays ›
Gene Symbol:
TPD52L1
Gene Name:
tumor protein D52-like 1
Gene Aliases:
D53
Chromosome Location:
Chr.6: 125153483 - 125264407 on Build GRCh38
UniGene ID:
Hs.591347
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14328
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPD52L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003395.2
NP_001003395.1
2, 3
466
NM_001003396.2
NP_001003396.1
2, 3
364
NM_001003397.2
NP_001003397.1
2, 3
364
NM_001292026.2
NP_001278955.1
2, 3
466
NM_001300994.2
NP_001287923.1
2, 3
364
NM_001318903.1
NP_001305832.1
2, 3
364
NM_001318907.1
NP_001305836.1
2, 3
261
NM_003287.3
NP_003278.1
2, 3
364
XM_005267122.2
XP_005267179.1
2, 3
145
XM_006715557.3
XP_006715620.1
2, 3
610
XM_017011237.1
XP_016866726.1
2, 3
610
XM_017011238.1
XP_016866727.1
2, 3
610
XM_017011239.1
XP_016866728.1
2, 3
466
XM_017011240.1
XP_016866729.1
2, 3
144
GenBank mRNA
AF004427.1
-
2, 3
181
AF208012.1
-
2, 3
180
AK124817.1
-
2, 3
466
AK291866.1
-
2, 3
328
AK291872.1
-
2, 3
198
AK291887.1
-
2, 3
159
BC002375.2
-
2, 3
254
2822877
BE409865.1
-
2, 3
392
BG770463.1
-
2, 3
262
BM048527.1
-
2, 3
384
BM546629.1
-
2, 3
237
BP334548.1
-
2, 3
280
BQ641312.1
-
2, 3
243
CB114736.1
-
2, 3
364
U44427.1
-
2, 3
321
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
positive regulation of MAP kinase activity
positive regulation of JNK cascade
positive regulation of apoptotic signaling pathway
protein binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-ffb8c786-kqswr:80/100.66.130.62:80.
git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0