Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100942.1 | NP_001094412.1 | 3 | 158 | ||
GenBank mRNA | BC161800.1 | - | 3 | 201 | 9026464 | |
EF535528.1 | - | 3 | 158 | |||
FQ217734.1 | - | 2 | 149 | |||
FQ221470.1 | - | 3 | 179 | |||
FQ224264.1 | - | 3 | 195 |
Gene Symbol: LOC100911238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248840.1 | XP_006248902.1 | 3 | 245 |