Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001371.1 | NP_001001371.1 | 1 | 799 |
Gene Symbol: LOC103692758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765057.1 | XP_008763279.1 | 1 | 1146 |
Gene Symbol: Olr1029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001366.1 | NP_001001366.1 | 1 | 799 |
Gene Symbol: Olr1058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000066.1 | NP_001000066.1 | 1 | 799 |
Gene Symbol: Olr956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001381.1 | NP_001001381.1 | 1 | 799 |