Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103693776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770394.1 | XP_008768616.1 | 2, 3 | 4192 | ||
XM_008770395.1 | XP_008768617.1 | 1, 2 | 118 | |||
XM_008770396.1 | XP_008768618.1 | 2, 3 | 4192 | |||
XM_008770397.1 | XP_008768619.1 | 2, 3 | 4192 | |||
XM_008770398.1 | XP_008768620.1 | 2, 3 | 4192 | |||
XM_008770399.1 | XP_008768621.1 | 2, 3 | 4192 | |||
XM_008770400.1 | XP_008768622.1 | 2, 3 | 4192 | |||
XM_008770404.1 | XP_008768626.1 | 2, 3 | 4192 |
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100507.1 | NP_001093977.1 | 680 | |||
XM_006251439.2 | XP_006251501.1 | 4192 | ||||
XM_006251440.2 | XP_006251502.1 | 4192 | ||||
XM_008770262.1 | XP_008768484.1 | 4192 | ||||
XM_008770263.1 | XP_008768485.1 | 118 | ||||
XM_008770264.1 | XP_008768486.1 | 4192 | ||||
XM_008770265.1 | XP_008768487.1 | 4192 | ||||
XM_008770269.1 | XP_008768491.1 | 4192 |