Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203366.2 | NP_976242.2 | 12 | 1469 | ||
NM_203367.2 | NP_976243.2 | 12 | 1637 | |||
NM_203368.2 | NP_976244.2 | 11 | 1307 | |||
NM_203369.2 | NP_976245.2 | 12 | 1556 | |||
XM_006254150.2 | XP_006254212.1 | 14 | 1867 | |||
XM_006254151.2 | XP_006254213.1 | 14 | 1920 | |||
XM_006254152.2 | XP_006254214.1 | 14 | 1779 | |||
XM_006254154.2 | XP_006254216.1 | 13 | 1822 | |||
XM_006254155.2 | XP_006254217.1 | 13 | 1874 | |||
XM_008771704.1 | XP_008769926.1 | 14 | 1917 | |||
XM_008771705.1 | XP_008769927.1 | 14 | 1750 | |||
XM_008771706.1 | XP_008769928.1 | 12 | 1610 | |||
XM_008771707.1 | XP_008769929.1 | 12 | 1712 | |||
XM_008771708.1 | XP_008769930.1 | 13 | 1704 | |||
XM_008771709.1 | XP_008769931.1 | 13 | 1702 | |||
XM_008771710.1 | XP_008769932.1 | 12 | 1542 | |||
XM_008771711.1 | XP_008769933.1 | 12 | 1540 | |||
GenBank mRNA | AB162617.1 | - | 11 | 1330 | ||
AB162618.1 | - | 11 | 1498 | |||
AB162619.1 | - | 10 | 1336 | |||
AB162620.1 | - | 10 | 1168 | |||
BC065308.1 | - | 12 | 1553 | 5599817 |