Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006969.1 | NP_001006970.1 | 8 | 1325 | ||
XM_006229021.1 | XP_006229083.1 | 8 | 1927 | |||
XM_006229022.2 | XP_006229084.1 | 8 | 1887 | |||
XM_006229023.2 | XP_006229085.1 | 8 | 1446 | |||
XM_006229024.2 | XP_006229086.1 | 8 | 1314 | |||
XM_008759337.1 | XP_008757559.1 | 7 | 1275 | |||
GenBank mRNA | BC083776.1 | - | 8 | 1325 | 7193483 | |
FQ218848.1 | - | 8 | 1276 |