Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013896.1 | NP_001013918.1 | 14 | 1384 | ||
GenBank mRNA | BC085115.1 | - | 14 | 1384 | 7106476 |