Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1686 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001373.1 | NP_001001373.1 | 1 | 392 |
Gene Symbol: LOC100910479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255857.2 | XP_006255919.1 | 725 |
Gene Symbol: Olr1702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001374.1 | NP_001001374.1 | 1 | 392 |