Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212462.2 | NP_997627.1 | 1 | 83 | ||
XM_006256048.2 | XP_006256110.1 | 2 | 328 | |||
XM_006256050.2 | XP_006256112.1 | 2 | 328 | |||
XM_006256051.2 | XP_006256113.1 | 2 | 327 | |||
XM_006256052.2 | XP_006256114.1 | 2 | 328 | |||
XM_006256053.2 | XP_006256115.1 | 2 | 328 |