Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008885.2 | NP_001008885.1 | 1 | 485 | ||
NM_001164744.1 | NP_001158216.1 | 2 | 226 | |||
GenBank mRNA | BC079141.1 | - | 1 | 485 | 7127902 | |
DY547565.1 | - | 2 | 226 | |||
FQ212335.1 | - | 2 | 193 | |||
FQ212833.1 | - | 1 | 513 |