Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam26f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024976.1 | NP_001020147.1 | 2 | 647 | ||
XM_008772880.1 | XP_008771102.1 | 2 | 536 | |||
GenBank mRNA | BC093379.1 | - | 2 | 647 | 7388112 |