Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037185.2 | NP_001032262.2 | 6 | 1106 | ||
GenBank mRNA | BC107944.1 | - | 6 | 880 | ||
FQ226690.1 | - | 4 | 510 | |||
FQ227460.1 | - | 3 | 369 | |||
FQ230642.1 | - | 3 | 427 | |||
FQ233743.1 | - | 4 | 510 |