Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNNC2 SIRNA Assays ›
          Gene Symbol:
          Tnnc2
          Gene Name:
          troponin C type 2 (fast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 161243747 - 161244831 on Build Rnor_6.0
          UniGene ID:
          Rn.43529
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s150097

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tnnc2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037351.1 NP_001032428.1 3 217
          GenBank mRNA DQ273679.1 - 3 217
          FQ215069.1 - 281
          FQ216869.1 - 281
          FQ216875.1 - 311
          FQ216940.1 - 273
          FQ216956.1 - 294
          FQ217047.1 - 281
          FQ217106.1 - 310
          FQ217137.1 - 281
          FQ217169.1 - 281
          FQ217172.1 - 281
          FQ217195.1 - 281
          FQ217236.1 - 292
          FQ217299.1 - 281
          FQ217371.1 - 281
          FQ217378.1 - 281
          FQ217706.1 - 281
          FQ217788.1 - 281
          FQ217840.1 - 281
          FQ217842.1 - 281
          FQ217896.1 - 280
          FQ217902.1 - 281
          FQ217984.1 - 281
          FQ218010.1 - 281
          FQ218048.1 - 311
          FQ218061.1 - 281
          FQ223543.1 - 281
          FQ223572.1 - 281
          FQ223583.1 - 281
          FQ223607.1 - 281
          FQ223623.1 - 281
          FQ223636.1 - 281
          FQ223643.1 - 281
          FQ223690.1 - 281
          FQ223692.1 - 281
          FQ223711.1 - 281
          FQ223713.1 - 281
          FQ223719.1 - 281
          FQ223773.1 - 293
          FQ223816.1 - 302
          FQ223825.1 - 281
          FQ223832.1 - 281
          FQ223835.1 - 311
          FQ223976.1 - 281
          FQ223986.1 - 281
          FQ224079.1 - 281
          FQ224114.1 - 310
          FQ224128.1 - 281
          FQ224223.1 - 281
          FQ224258.1 - 281
          FQ224340.1 - 281
          FQ224376.1 - 296
          FQ224378.1 - 281
          FQ224387.1 - 281
          FQ224450.1 - 281
          FQ224462.1 - 281
          FQ224599.1 - 281
          FQ224648.1 - 281
          FQ224672.1 - 281
          FQ224735.1 - 282
          FQ224749.1 - 281
          FQ224795.1 - 281
          FQ224811.1 - 281
          FQ224822.1 - 281
          FQ224827.1 - 278

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          actin binding
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0