Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013924.1 | NP_001013946.1 | 8 | 1178 | ||
XM_006235817.2 | XP_006235879.1 | 8 | 1604 | |||
XM_006235818.2 | XP_006235880.1 | 8 | 1224 | |||
XM_006235820.2 | XP_006235882.1 | 8 | 1232 | |||
XM_008762485.1 | XP_008760707.1 | 7 | 2738 | |||
XM_008762486.1 | XP_008760708.1 | 8 | 1608 | |||
XM_008762487.1 | XP_008760709.1 | 9 | 1325 | |||
XM_008762488.1 | XP_008760710.1 | 9 | 1348 | |||
XM_008762489.1 | XP_008760711.1 | 8 | 1247 | |||
XM_008762490.1 | XP_008760712.1 | 9 | 1304 | |||
XM_008762491.1 | XP_008760713.1 | 8 | 1228 | |||
XM_008762492.1 | XP_008760714.1 | 6 | 2519 | |||
XM_008762493.1 | XP_008760715.1 | 8 | 1255 | |||
XM_008762494.1 | XP_008760716.1 | 7 | 2731 | |||
XM_008762495.1 | XP_008760717.1 | 6 | 2599 | |||
GenBank mRNA | BC083707.1 | - | 8 | 1178 | 7190542 |