Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106613.2 | NP_001100083.2 | 6 | 684 | ||
XM_006236962.2 | XP_006237024.1 | 6 | 758 | |||
GenBank mRNA | BC167050.1 | - | 6 | 602 | 7114402 | |
EV776660.1 | - | 3 | 163 |