Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106689.1 | NP_001100159.1 | 2 | 352 | ||
GenBank mRNA | BC166512.1 | - | 2 | 308 | 7128768 |