Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005903.1 | NP_001005903.1 | 5 | 478 | ||
XM_006239171.2 | XP_006239233.1 | 6 | 1530 | |||
XM_006239172.2 | XP_006239234.1 | 6 | 1527 | |||
XM_006239174.2 | XP_006239236.1 | 5 | 1457 | |||
XM_006239175.2 | XP_006239237.1 | 5 | 1453 | |||
XM_006239177.2 | XP_006239239.1 | 5 | 507 | |||
GenBank mRNA | BC083861.1 | - | 5 | 478 | 7126848 |