Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106778.1 | NP_001100248.1 | 19 | 2295 | ||
XM_008765227.1 | XP_008763449.1 | 17 | 7551 | |||
XM_008765228.1 | XP_008763450.1 | 17 | 1586 | |||
XM_008765229.1 | XP_008763451.1 | 16 | 7506 | |||
XM_008765230.1 | XP_008763452.1 | 16 | 1481 | |||
XM_008765231.1 | XP_008763453.1 | 21 | 3219 |