Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPL3 SIRNA Assays ›
          Gene Symbol:
          Rpl3
          Gene Name:
          ribosomal protein L3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 121297365 - 121302739 on Build Rnor_6.0
          UniGene ID:
          Rn.107726
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s152674

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpl3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198753.2 NP_942048.1 3 361
          GenBank mRNA BC058494.1 - 3 364 6887923
          FQ209477.1 - 3 360
          FQ211542.1 - 3 361
          FQ211764.1 - 3 361
          FQ212671.1 - 3 361
          FQ213017.1 - 3 361
          FQ213135.1 - 3 363
          FQ214177.1 - 3 361
          FQ214350.1 - 3 361
          FQ214687.1 - 3 358
          FQ215305.1 - 3 361
          FQ218147.1 - 3 361
          FQ219083.1 - 3 361
          FQ219302.1 - 3 361
          FQ219523.1 - 3 360
          FQ220029.1 - 3 360
          FQ220099.1 - 3 361
          FQ220378.1 - 3 361
          FQ220408.1 - 3 357
          FQ220423.1 - 3 360
          FQ220830.1 - 3 361
          FQ222720.1 - 3 361
          FQ222993.1 - 3 361
          FQ223514.1 - 3 361
          FQ225290.1 - 3 361
          FQ225405.1 - 3 361
          FQ225635.1 - 3 361
          FQ226331.1 - 3 361
          FQ227897.1 - 3 361
          FQ228916.1 - 3 361
          FQ229153.1 - 3 361
          FQ229505.1 - 3 361
          FQ229795.1 - 3 357
          FQ229869.1 - 3 361
          FQ229906.1 - 3 361
          FQ230150.1 - 3 361
          FQ231551.1 - 3 357
          FQ232038.1 - 3 361
          FQ232101.1 - 3 361
          FQ232351.1 - 1 103
          FQ233196.1 - 3 361
          FQ234897.1 - 3 361
          FQ235165.1 - 3 361
          X62166.1 - 3 365

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_013762, AK151596, AK168128, AK088993, AK169321, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          cellular response to interleukin-4
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0