Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212516.2 | NP_997681.1 | 2 | 229 | ||
GenBank mRNA | BC058497.1 | - | 2 | 230 | 6918545 | |
FQ210248.1 | - | 2 | 219 | |||
FQ211867.1 | - | 2 | 226 | |||
FQ214576.1 | - | 225 | ||||
FQ217032.1 | - | 2 | 212 | |||
FQ217139.1 | - | 2 | 245 | |||
FQ217410.1 | - | 2 | 246 | |||
FQ217906.1 | - | 2 | 218 | |||
FQ218008.1 | - | 2 | 218 | |||
FQ221909.1 | - | 2 | 258 | |||
FQ223878.1 | - | 2 | 219 | |||
FQ223920.1 | - | 2 | 225 | |||
FQ224004.1 | - | 2 | 219 | |||
FQ224067.1 | - | 2 | 219 | |||
FQ224620.1 | - | 2 | 219 | |||
FQ224644.1 | - | 2 | 225 |