Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013108.3 | NP_001013126.2 | 3 | 769 | ||
GenBank mRNA | BC083881.1 | - | 3 | 769 | 7131418 |