Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013109.1 | NP_001013127.1 | 1 | 176 | ||
XM_006243306.2 | XP_006243368.1 | 5 | 693 | |||
XM_006243307.1 | XP_006243369.1 | 2 | 170 | |||
XM_006243308.2 | XP_006243370.1 | 2 | 1372 | |||
XM_006243309.2 | XP_006243371.1 | 2 | 1067 | |||
XM_008766241.1 | XP_008764463.1 | 5 | 703 | |||
GenBank mRNA | BC091410.1 | - | 1 | 176 | 7325495 |