Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004250.2 | NP_001004250.1 | 12 | 1348 | ||
GenBank mRNA | BC078923.1 | - | 12 | 1350 | 7106265 | |
FQ210217.1 | - | 5 | 467 | |||
FQ213124.1 | - | 12 | 1359 | |||
FQ215592.1 | - | 8 | 832 | |||
FQ216746.1 | - | 12 | 1352 | |||
FQ224157.1 | - | 12 | 1356 | |||
FQ229052.1 | - | 12 | 1375 | |||
FQ231552.1 | - | 12 | 1372 |