Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127531.2 | NP_001121003.1 | 4 | 454 | ||
XM_006256860.2 | XP_006256922.1 | 4 | 526 | |||
XM_006256862.2 | XP_006256924.1 | 4 | 525 | |||
XM_008773167.1 | XP_008771389.1 | 4 | 526 | |||
GenBank mRNA | BC166480.1 | - | 4 | 454 | 9102341 |