Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008335.1 | NP_001008336.1 | 10 | 1061 | ||
XM_006248534.2 | XP_006248596.1 | 10 | 1077 | |||
XM_006248535.2 | XP_006248597.1 | 10 | 1077 | |||
GenBank mRNA | BC085859.1 | - | 10 | 1061 | 7190333 | |
FQ215479.1 | - | 3 | 214 |