Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107277.1 | NP_001100747.1 | 6 | 898 | ||
XM_006252273.2 | XP_006252335.1 | 5 | 817 |