Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt8d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007683.1 | NP_001007684.1 | 9 | 1041 | ||
GenBank mRNA | AY325256.1 | - | 9 | 988 | ||
BC079066.1 | - | 9 | 1041 | 7121464 |