Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025675.1 | NP_001020846.1 | 3, 4 | 302 | ||
GenBank mRNA | BC097977.1 | - | 3, 4 | 302 | 7442728 |