Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC108823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012353.1 | NP_001012353.1 | 2 | 1281 | ||
GenBank mRNA | BC089836.1 | - | 2 | 1281 | 7326302 |
Gene Symbol: LOC102555392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254757.2 | XP_006254819.1 | 1163 |
Gene Symbol: LOC103694226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772196.1 | XP_008770418.1 | 1 | 1163 |