Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107430.1 | NP_001100900.1 | 15 | 2043 | ||
XM_006255596.2 | XP_006255658.2 | 17 | 2298 | |||
XM_008772514.1 | XP_008770736.1 | 16 | 2214 |